Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Prpsap1
Entrez Gene ID
67763
Gene Name
phosphoribosyl pyrophosphate synthetase-associated protein 1
Gene Aliases
5730409F23Rik, PAP39
Chromosome Location
Chr.11: 116470816 - 116494652 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_026364.1 NP_080640.1 5 - 6 710 82
GenBank mRNA AK011290.1 - 5 - 6 680 82
AK208020.1 - 2 - 3 237 82
AK218181.1 - 5 - 6 342 82
BC029621.1 - 5 - 6 701 82

View Full Product Details