Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mospd2
Entrez Gene ID
76763
Gene Name
motile sperm domain containing 2
Gene Aliases
2410013I23Rik
Chromosome Location
Chr.X: 164936169 - 164980375 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001290523.1 NP_001277452.1 8 - 9 853 125
NM_001290524.1 NP_001277453.1 8 - 9 853 125
NM_029730.4 NP_084006.2 8 - 9 853 125
GenBank mRNA AK010480.1 - 8 - 9 775 125
AK031405.1 - 8 - 9 778 125
AK035985.1 - 8 - 9 813 125
BC026425.1 - 8 - 9 779 125

View Full Product Details