The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001177956.1 | NP_001171427.1 | 7 - 8 | 1304 | 107 |
NM_001177957.1 | NP_001171428.1 | 7 - 8 | 1304 | 107 | |
NM_001177959.1 | NP_001171430.1 | 6 - 7 | 1184 | 107 | |
NM_001177960.1 | NP_001171431.1 | 6 - 7 | 1184 | 107 | |
NM_001177961.1 | NP_001171432.1 | 6 - 7 | 849 | 107 | |
NM_001177962.1 | NP_001171433.1 | 6 - 7 | 849 | 107 | |
GenBank mRNA | AF254872.1 | - | 6 - 7 | 1045 | 107 |
AF254873.1 | - | 6 - 7 | 1045 | 107 | |
AF254874.1 | - | 7 - 8 | 1165 | 107 | |
AF254875.1 | - | 7 - 8 | 1165 | 107 | |
AF254876.1 | - | 6 - 7 | 820 | 107 | |
AF254877.1 | - | 6 - 7 | 820 | 107 | |
AK132778.1 | - | 6 - 7 | 836 | 107 | |
BC003912.1 | - | 5 - 6 | 632 | 107 | |
BC025165.1 | - | 6 - 7 | 810 | 107 |