The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001168568.1 | NP_001162040.1 | 18 - 19 | 2035 | 82 |
NM_001168569.1 | NP_001162041.1 | 18 - 19 | 2092 | 82 | |
NM_001168571.1 | NP_001162043.1 | 16 - 17 | 2014 | 82 | |
NM_018737.5 | NP_061207.1 | 18 - 19 | 2167 | 82 | |
NR_033143.1 | 18 - 19 | 2057 | 82 | ||
GenBank mRNA | AK032018.1 | - | 18 - 19 | 1894 | 82 |
AK052854.1 | - | 18 - 19 | 2083 | 82 | |
AK084595.1 | - | 18 - 19 | 1964 | 82 | |
AK085381.1 | - | 18 - 19 | 1943 | 82 | |
AK150966.1 | - | 18 - 19 | 2034 | 82 | |
AK151597.1 | - | 18 - 19 | 2034 | 82 | |
AK164206.1 | - | 16 - 17 | 1841 | 82 | |
BC003257.1 | - | 18 - 19 | 1875 | 82 | |
EF410142.1 | - | - | 766 | 82 | |
U49385.1 | - | 18 - 19 | 2057 | 82 |