The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_183308.2 | NP_899131.1 | 1 - 2 | 163 | 117 |
GenBank mRNA | AK007350.1 | - | 1 - 2 | 131 | 117 |
AK146311.1 | - | 1 - 2 | 131 | 117 | |
AK152351.1 | - | 1 - 2 | 114 | 117 | |
AK171926.1 | - | 1 - 2 | 121 | 117 | |
BC021887.1 | - | 1 - 2 | 120 | 117 | |
BC037140.1 | - | 1 - 2 | 105 | 117 | |
BC055896.1 | - | 1 - 2 | 163 | 117 | |
BC062200.1 | - | 1 - 2 | 154 | 117 |