The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_133692.2 | NP_598453.1 | 7 - 8 | 798 | 118 |
GenBank mRNA | AF294329.1 | - | 7 - 8 | 809 | 118 |
AK031778.1 | - | 7 - 8 | 800 | 118 | |
AK088385.1 | - | 7 - 8 | 791 | 118 | |
AK162240.1 | - | 2 - 3 | 127 | 118 | |
AK168179.1 | - | 7 - 8 | 786 | 118 | |
BC055325.1 | - | 7 - 8 | 801 | 118 |