Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dpys
Entrez Gene ID
64705
Gene Name
dihydropyrimidinase
Gene Aliases
1200017I10Rik, 1300004I01Rik, DHP, DHPase
Chromosome Location
Chr.15: 39768485 - 39857470 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001164466.1 NP_001157938.1 7 - 8 1340 85
NM_022722.3 NP_073559.3 7 - 8 1340 85
GenBank mRNA AF227731.1 - 7 - 8 1258 85
AF249296.1 - 7 - 8 1327 85
AK004822.1 - 7 - 8 1340 85
AK004899.2 - 7 - 8 1337 85
BC029718.1 - 5 - 6 679 85
BC037086.1 - 7 - 8 1309 85

View Full Product Details