Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pop1
Entrez Gene ID
67724
Gene Name
processing of precursor 1, ribonuclease P/MRP family, (S. cerevisiae)
Gene Aliases
4932434G09Rik
Chromosome Location
Chr.15: 34495311 - 34530653 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_026340.3 NP_080616.1 15 - 16 2512 108
NM_152894.2 NP_690854.1 16 - 17 2602 108
GenBank mRNA AK016547.1 - 15 - 16 2499 108
BC034742.1 - 16 - 17 2581 108

View Full Product Details