Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Lrrc27
Entrez Gene ID
76612
Gene Name
leucine rich repeat containing 27
Gene Aliases
1700071K18Rik, 2310044E02Rik
Chromosome Location
Chr.7: 139212869 - 139242979 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001143755.1 NP_001137227.1 4 - 5 649 113
NM_027164.1 NP_081440.1 3 - 4 694 113
GenBank mRNA AK006931.1 - 4 - 5 654 113
BC050816.1 - 3 - 4 694 113

View Full Product Details