Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ero1lb
Entrez Gene ID
67475
Gene Name
ERO1-like beta (S. cerevisiae)
Gene Aliases
1300013B24Rik, 1700065B09Rik, AI447560, ero1-beta
Chromosome Location
Chr.13: 12565809 - 12609530 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_026184.2 NP_080460.2 12 - 13 1241 74
GenBank mRNA AF489856.1 - 12 - 13 1047 74
AK004981.1 - - 1218 74
AK006886.1 - 12 - 13 1101 74
AK011964.1 - - 114 74
AK041951.1 - 12 - 13 1217 74
AK160664.1 - 12 - 13 1257 74
BC058721.1 - 12 - 13 1241 74
BC111900.1 - 12 - 13 1211 74
BC112426.1 - 12 - 13 1211 74
BC145888.1 - 12 - 13 1192 74
BC145890.1 - 12 - 13 1192 74

View Full Product Details