Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Sept7
Entrez Gene ID
235072
Gene Name
septin 7
Gene Aliases
Cdc10, E430034N22
Chromosome Location
Chr.9: 25251090 - 25308571 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205367.1 NP_001192296.1 7 - 8 916 125
NM_009859.4 NP_033989.2 8 - 9 919 125
GenBank mRNA AK088997.1 - 8 - 9 706 125
AK144127.1 - 8 - 9 708 125
AK220447.1 - 7 - 8 689 125
BC058587.1 - 7 - 8 720 125

View Full Product Details