Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Hmgn3
Entrez Gene ID
94353
Gene Name
high mobility group nucleosomal binding domain 3
Gene Aliases
1110002A15Rik, 6330514M13Rik, BB071015, TRIP7
Chromosome Location
Chr.9: 83109942 - 83146734 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_026122.4 NP_080398.1 1 - 2 179 61
NM_175074.2 NP_778249.1 1 - 2 179 61
GenBank mRNA AK002970.1 - 1 - 2 98 61
AK003272.1 - 1 - 2 127 61
AK153223.1 - 1 - 2 139 61
BC005693.1 - 1 - 2 115 61

View Full Product Details