Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tmod2
Entrez Gene ID
50876
Gene Name
tropomodulin 2
Gene Aliases
N-Tmod, NTMOD
Chromosome Location
Chr.9: 75565621 - 75611348 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001038710.1 NP_001033799.1 7 - 8 1165 65
NM_016711.3 NP_057920.2 7 - 8 1101 65
GenBank mRNA AF177170.1 - 7 - 8 1050 65
AF237629.1 - 8 - 9 1009 65
AK017072.1 - 7 - 8 1149 65
AK076562.1 - 7 - 8 1115 65
AK079077.1 - 7 - 8 945 65
AK147581.1 - 7 - 8 1097 65
AK164178.1 - 7 - 8 947 65
BC052227.1 - 7 - 8 1165 65
BC061124.1 - 7 - 8 931 65

View Full Product Details