The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_011220.2 | NP_035350.1 | 5 - 6 | 310 | 69 |
GenBank mRNA | AF043225.1 | - | 5 - 6 | 325 | 69 |
AK002614.1 | - | 5 - 6 | 320 | 69 | |
AK146987.1 | - | 5 - 6 | 321 | 69 | |
AK153959.1 | - | 3 - 4 | 2393 | 69 | |
BC029013.1 | - | 5 - 6 | 310 | 69 |