Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Impg1
Entrez Gene ID
63859
Gene Name
interphotoreceptor matrix proteoglycan 1
Gene Aliases
A930015H12Rik, IMP150, SPACR
Chromosome Location
Chr.9: 80308757 - 80465654 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_022016.3 NP_071299.3 8 - 9 1028 138
GenBank mRNA AF229929.1 - 8 - 9 939 138
AF266478.1 - 8 - 9 1074 138
BC022970.1 - 8 - 9 1016 138

View Full Product Details