The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_010295.2 | NP_034425.1 | 1 - 2 | 424 | 60 |
GenBank mRNA | AK144339.1 | - | 1 - 2 | 425 | 60 |
AK160876.1 | - | 1 - 2 | 184 | 60 | |
AK169676.1 | - | 1 - 2 | 397 | 60 | |
BC019374.1 | - | 1 - 2 | 181 | 60 | |
U85414.1 | - | 1 - 2 | 149 | 60 | |
U85498.1 | - | 1 - 2 | 149 | 60 |