Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ccdc82
Entrez Gene ID
66396
Gene Name
coiled-coil domain containing 82
Gene Aliases
2310043N13Rik
Chromosome Location
Chr.9: 13246979 - 13292353 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK009932.1 - 1 - 2 34 128
AK166406.1 - 1 - 2 60 128
BC098496.1 - 1 - 2 47 128

View Full Product Details