Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Sirt2
Entrez Gene ID
64383
Gene Name
sirtuin 2
Gene Aliases
5730427M03Rik, SIR2L2, Sir2l
Chromosome Location
Chr.7: 28766752 - 28788665 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001122765.1 NP_001116237.1 7 - 8 556 75
NM_001122766.1 NP_001116238.1 5 - 6 393 75
NM_022432.4 NP_071877.3 8 - 9 603 75
GenBank mRNA AB221570.1 - 8 - 9 508 75
AF299337.1 - 8 - 9 588 75
AK014042.1 - 7 - 8 527 75
AK144431.1 - 7 - 8 500 76
AK146412.1 - 7 - 8 536 75
BC021439.1 - 8 - 9 607 75
KF032392.1 - 8 - 9 545 75

View Full Product Details