Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pou2f2
Entrez Gene ID
18987
Gene Name
POU domain, class 2, transcription factor 2
Gene Aliases
Oct-2, Oct2a, Oct2b, Otf-2, Otf2
Chromosome Location
Chr.7: 25091114 - 25179992 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001163554.1 NP_001157026.1 1 - 2 31 83
NM_001163555.1 NP_001157027.1 1 - 2 31 83
NM_001163556.1 NP_001157028.1 1 - 2 31 83
NM_011138.2 NP_035268.2 1 - 2 31 83
GenBank mRNA AK156322.1 - 1 - 2 44 83
AY746973.1 - 1 - 2 59 83
BC050258.1 - 1 - 2 44 83
BC104488.1 - 1 - 2 31 83
BC105647.1 - 1 - 2 31 83
BC105920.1 - 1 - 2 31 83
BC105921.1 - 1 - 2 31 83
X53654.1 - - 239 83
X57936.1 - 1 - 2 31 83
X57937.1 - 1 - 2 31 83
X57938.1 - 1 - 2 31 83
X57939.1 - 1 - 2 31 83
X57940.1 - 1 - 2 31 83
X57941.1 - 1 - 2 31 83

View Full Product Details