Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pcbp3
Entrez Gene ID
59093
Gene Name
poly(rC) binding protein 3
Gene Aliases
AlphaCP-3
Chromosome Location
Chr.10: 76761854 - 76961953 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_021568.2 NP_067543.2 9 - 10 1023 67
GenBank mRNA AF176327.1 - 9 - 10 864 67
AK034811.1 - 12 - 13 1402 67
AK079564.1 - 7 - 8 1065 67
BC042440.1 - 7 - 8 658 67

View Full Product Details