Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mmp11
Entrez Gene ID
17385
Gene Name
matrix metallopeptidase 11
Gene Aliases
MMP-11, SL-3, ST3, Stmy3
Chromosome Location
Chr.10: 75923221 - 75932520 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK142169.1 - 6 - 7 1111 64
AK165778.1 - 6 - 7 1114 64
BC052854.1 - 6 - 7 1113 64
Z12604.1 - 6 - 7 1099 64

View Full Product Details