The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_011179.3 | NP_035309.3 | 7 - 9 | 895 | 81 |
GenBank mRNA | AK148545.1 | - | 7 - 9 | 811 | 81 |
S36200.1 | - | 7 - 9 | 785 | 81 | |
S71616.1 | - | 7 - 9 | 825 | 81 |