The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_027829.3 | NP_082105.3 | 8 - 9 | 607 | 109 |
GenBank mRNA | BF682661.1 | - | 8 - 9 | 541 | 109 |
CB055593.1 | - | 7 - 8 | 607 | 109 |