Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Omt2a
Entrez Gene ID
18379
Gene Name
oocyte maturation, alpha
Gene Aliases
OM2a
Chromosome Location
Chr.9: 78311972 - 78314797 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001111286.2 NP_001104756.1 1 - 2 726 115
GenBank mRNA AK135846.1 - 1 - 2 152 115
AK139546.1 - 1 - 2 694 115
DV647767.1 - 1 - 2 158 115
DV648064.1 - 1 - 2 170 115
S81934.1 - 2 - 3 262 115

View Full Product Details