Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cdkl5
Entrez Gene ID
382253
Gene Name
cyclin-dependent kinase-like 5
Gene Aliases
BC038161, Stk9
Chromosome Location
Chr.X: 160784308 - 160994681 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001024624.2 NP_001019795.1 10 - 11 1080 102
GenBank mRNA AK052380.1 - 10 - 11 1077 102
AK139220.1 - 10 - 11 1052 102

View Full Product Details