The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_139143.3 | NP_631882.2 | 1 - 2 | 534 | 128 |
GenBank mRNA | AK028976.1 | - | 1 - 2 | 493 | 128 |
BC055012.1 | - | 1 - 2 | 204 | 128 | |
CK619600.1 | - | 1 - 2 | 534 | 128 |