Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Nrg2
Entrez Gene ID
100042150
Gene Name
neuregulin 2
Gene Aliases
Don1, NTAK
Chromosome Location
Chr.18: 36016758 - 36199047 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001167891.1 NP_001161363.1 6 - 7 1322 65

View Full Product Details