Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Slc26a10
Entrez Gene ID
216441
Gene Name
solute carrier family 26, member 10
Gene Aliases
C78409, D630010I07
Chromosome Location
Chr.10: 127172313 - 127180709 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_177615.3 NP_808283.2 16 - 17 1962 59
GenBank mRNA AK052641.1 - - 2316 59
BC089587.1 - 15 - 16 2314 59

View Full Product Details