The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001048207.1 | NP_001041672.1 | 2 - 3 | 299 | 58 |
GenBank mRNA | AK002769.1 | - | 2 - 3 | 129 | 58 |
AK082930.1 | - | 2 - 3 | 213 | 58 | |
AK152607.1 | - | 2 - 3 | 229 | 58 | |
AK159730.1 | - | 2 - 3 | 233 | 58 | |
AK162472.1 | - | 1 - 2 | 33 | 58 | |
AY234183.1 | - | 2 - 3 | 299 | 58 | |
BC027408.1 | - | 2 - 3 | 239 | 58 |