Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Prrt1
Entrez Gene ID
260297
Gene Name
proline-rich transmembrane protein 1
Gene Aliases
Bat-4, Bat4, D17H6S54E, DSPD1, G5b, NG5, ORF31
Chromosome Location
Chr.17: 34629569 - 34633134 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_030890.1 NP_112152.1 3 - 4 747 56
GenBank mRNA BC115512.2 - 3 - 4 550 56
BC115513.2 - 3 - 4 550 56
BC128489.1 - 4 - 5 416 56

View Full Product Details