The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001167757.1 | NP_001161229.1 | 3 - 4 | 703 | 177 |
NM_029202.2 | NP_083478.2 | 3 - 4 | 700 | 177 | |
GenBank mRNA | AK015948.1 | - | 3 - 4 | 644 | 177 |
AK077080.1 | - | 3 - 4 | 672 | 177 | |
BC049731.1 | - | 3 - 4 | 700 | 177 |