Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Nol4
Entrez Gene ID
319211
Gene Name
nucleolar protein 4
Gene Aliases
1700013J13Rik, 4930568N03Rik, Gm1262
Chromosome Location
Chr.18: 22693152 - 23041675 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001161483.1 NP_001154955.1 2 - 3 252 63
NM_199024.2 NP_950189.2 2 - 3 2022 63
GenBank mRNA AK031008.1 - 2 - 3 2022 63
BC056377.1 - 2 - 3 245 63

View Full Product Details