Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Prim1
Entrez Gene ID
19075
Gene Name
DNA primase, p49 subunit
Gene Aliases
AI324982
Chromosome Location
Chr.10: 128015215 - 128030030 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_008921.2 NP_032947.1 2 - 3 296 98
GenBank mRNA AK088557.1 - 2 - 3 316 98
AK089063.1 - 2 - 3 321 98
AK133878.1 - 2 - 3 293 98
AK160831.1 - 2 - 3 316 98
AK169632.1 - 2 - 3 293 98
BC089544.1 - 2 - 3 335 98
D13544.1 - 2 - 3 296 98
J04620.1 - 2 - 3 355 98

View Full Product Details