Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Prl7d1
- Entrez Gene ID
-
18814
- Gene Name
- prolactin family 7, subfamily d, member 1
- Gene Aliases
-
PLF-RP, PRP, Plfr
- Chromosome Location
-
Chr.13: 27708998 - 27716748 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK005421.1 |
- |
1 - 2 |
101 |
75 |
View Full Product Details