The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_181680.1 | NP_859008.1 | 9 - 10 | 1373 | 79 |
GenBank mRNA | AK034104.1 | - | - | 1316 | 79 |
AY192159.1 | - | 9 - 10 | 1373 | 79 |