Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Serpinh1
Entrez Gene ID
12406
Gene Name
serine (or cysteine) peptidase inhibitor, clade H, member 1
Gene Aliases
BERF-1, Cbp1, Cbp2, Hsp47, J6, Serpinh2, gp46
Chromosome Location
Chr.7: 99345375 - 99353239 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001111043.1 NP_001104513.1 1 - 2 187 81
GenBank mRNA AK077992.1 - 1 - 2 63 81
BC085143.1 - 1 - 2 60 81
J05609.1 - 1 - 2 61 81

View Full Product Details