Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Nupl2
Entrez Gene ID
231042
Gene Name
nucleoporin like 2
Gene Aliases
AV116043, CG1, NLP-1, Nlp1
Chromosome Location
Chr.5: 24164700 - 24184008 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_153092.4 NP_694732.3 5 - 6 726 175
GenBank mRNA AK077066.1 - 5 - 6 693 175
AK078478.1 - 5 - 6 727 175
BC033270.1 - 5 - 6 668 175

View Full Product Details