The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001146292.1 | NP_001139764.1 | 7 - 8 | 1307 | 67 |
NM_001146293.1 | NP_001139765.1 | 7 - 8 | 1310 | 67 | |
NM_001146294.1 | NP_001139766.1 | 7 - 8 | 1277 | 67 | |
NM_001174074.1 | NP_001167545.1 | 7 - 8 | 1307 | 67 | |
NM_133195.3 | NP_573458.2 | 7 - 8 | 1307 | 67 | |
GenBank mRNA | AF314173.1 | - | 4 - 5 | 393 | 67 |
AF515450.1 | - | 7 - 8 | 1045 | 67 | |
AK048427.1 | - | 7 - 8 | 1484 | 67 | |
AK051458.1 | - | 7 - 8 | 1028 | 67 | |
BC048405.1 | - | 7 - 8 | 1307 | 67 | |
BC052744.1 | - | 7 - 8 | 1310 | 67 |