Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Phrf1
Entrez Gene ID
101471
Gene Name
PHD and ring finger domains 1
Gene Aliases
AA673488
Chromosome Location
Chr.7: 141228263 - 141262751 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081118.1 NP_001074587.1 12 - 13 1588 137
GenBank mRNA BC052534.1 - 2 - 3 811 137
BC094566.1 - 1 - 2 63 137
BC138446.1 - 7 - 8 1234 137
BC145719.1 - 7 - 8 1234 137

View Full Product Details