Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Fastk
Entrez Gene ID
66587
Gene Name
Fas-activated serine/threonine kinase
Gene Aliases
0610011K02Rik, 610011K02Rik
Chromosome Location
Chr.5: 24441039 - 24445282 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_023229.2 NP_075718.2 8 - 9 1506 87
GenBank mRNA AF168682.1 - 7 - 8 1401 87
AK002552.1 - 8 - 9 1454 87
AK003940.1 - 5 - 6 749 87
AK038596.1 - 8 - 9 1458 87
AK052667.1 - 8 - 9 1451 87
AK139374.1 - 7 - 8 2226 87
AK151859.1 - 8 - 9 1458 87
AK152369.1 - 8 - 9 1507 87
AK159751.1 - 8 - 9 1458 87
AK164317.1 - - 1464 87
AK167346.1 - 7 - 8 2218 87
AK186668.1 - 1 - 2 154 87
BC013547.1 - 8 - 9 1423 87

View Full Product Details