The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_080467.3 | NP_536715.3 | 20 - 21 | 2689 | 115 |
GenBank mRNA | AB050903.1 | - | 19 - 20 | 2415 | 115 |
AF326316.1 | - | 20 - 21 | 2483 | 115 | |
BC046979.1 | - | 21 - 22 | 2711 | 115 |