Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Atp6v0a4
Entrez Gene ID
140494
Gene Name
ATPase, H+ transporting, lysosomal V0 subunit A4
Gene Aliases
Atp6n1b, a4
Chromosome Location
Chr.6: 38048483 - 38124586 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_080467.3 NP_536715.3 2 - 3 397 78
GenBank mRNA AB050903.1 - 1 - 2 123 78
AF326316.1 - 2 - 3 191 78
AF435090.1 - 2 - 3 385 78
AK128892.1 - 2 - 3 397 78
AK143790.1 - 2 - 3 398 78
AK165464.1 - 2 - 3 361 78
BC046979.1 - 3 - 4 419 78

View Full Product Details