The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_028957.2 | NP_083233.1 | 3 - 4 | 1340 | 127 |
GenBank mRNA | AK017112.1 | - | 3 - 4 | 1355 | 127 |
AK133387.1 | - | 3 - 4 | 1343 | 127 | |
BC145992.1 | - | 3 - 4 | 1273 | 127 | |
BC145994.1 | - | 3 - 4 | 1273 | 127 |