Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cdh12
Entrez Gene ID
215654
Gene Name
cadherin 12
Gene Aliases
Cdhb
Chromosome Location
Chr.15: 20448879 - 21597005 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001008420.2 NP_001008420.1 9 - 10 1718 79
GenBank mRNA BC086655.1 - 9 - 10 1718 79

View Full Product Details