Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Lat
Entrez Gene ID
16797
Gene Name
linker for activation of T cells
Gene Aliases
-
Chromosome Location
Chr.7: 126363827 - 126369589 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_010689.2 NP_034819.1 7 - 8 705 69
GenBank mRNA AF036907.1 - 7 - 8 705 69
AK134003.1 - 7 - 8 657 69
BC013337.1 - 7 - 8 682 69
BC052340.1 - 7 - 8 650 69

View Full Product Details