Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tac1
Entrez Gene ID
21333
Gene Name
tachykinin 1
Gene Aliases
4930528L02Rik, NK-1, NK1, Nkna, PPT-A, PPTA, SP
Chromosome Location
Chr.6: 7554776 - 7562978 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009311.2 NP_033337.1 3 - 4 315 77
GenBank mRNA BC117081.1 - 3 - 4 290 77
BC119122.1 - 3 - 4 290 77
D17584.1 - 3 - 4 315 77
M68908.1 - 2 - 3 111 77

View Full Product Details