Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Trbv1
Entrez Gene ID
621968
Gene Name
T cell receptor beta, variable 1
Gene Aliases
Gm6273, TCRBV2S1, Tcrb-V2
Chromosome Location
Chr.6: 40891296 - 40891885 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK138971.1 - - 123 80
M16680.1 - 1 - 2 54 80

View Full Product Details