The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_146861.1 | NP_667072.1 | 1 - 1 | 814 | 64 |
GenBank mRNA | BC116994.1 | - | 1 - 1 | 879 | 64 |
BC119365.1 | - | 1 - 1 | 879 | 64 |