Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fam71f2
Entrez Gene ID
245884
Gene Name
family with sequence similarity 71, member F2
Gene Aliases
AI507611, GARI
Chromosome Location
Chr.6: 29277539 - 29290684 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001101486.1 NP_001094956.1 1 - 2 282 83
GenBank mRNA AB473724.1 - 1 - 2 158 83
BC050974.1 - 1 - 2 282 83
BC151115.1 - 1 - 2 249 83
BC151117.1 - 1 - 2 249 83

View Full Product Details