The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_011752.2 | NP_035882.1 | 13 - 14 | 1378 | 85 |
GenBank mRNA | AB041612.1 | - | 13 - 14 | 1261 | 85 |
AK016551.1 | - | 7 - 8 | 545 | 85 | |
AK158995.1 | - | 13 - 14 | 1339 | 85 | |
AK167849.1 | - | 13 - 14 | 1249 | 85 | |
AK170372.1 | - | 4 - 5 | 2041 | 85 |